Dimethyl (4-methoxybenzyl)malonate 95%

Warning

H26331-03
AAH26331-03EA 73.62 USD
AAH26331-03
Dimethyl (4-methoxybenzyl)malonate 95%
Dimethyl (4-methoxybenzyl)malonate
Formula: C₁₃H₁₆O₅
MW: 252.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436113
CAS Number: 15378-09-3
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR