Diethyl (2,4-dichlorophenyl)malonate 95%

Supplier: Thermo Scientific Chemicals

Warning

H26330-ME H26330-27
AAH26330-27EA 272 USD
AAH26330-27 AAH26330-ME
Diethyl (2,4-dichlorophenyl)malonate 95%
Diethyl (2,4-dichlorophenyl)malonate
Formula: C₁₃H₁₄Cl₂O₄
MW: 305.16
Storage Temperature: Ambient
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR