1-Thien-2-ylpentan-1-one 97%

H26326-09 H26326-03
AAH26326-03EA 21.85 USD
AAH26326-03 AAH26326-09
1-Thien-2-ylpentan-1-one 97%
1-Thien-2-ylpentan-1-one
Formula: C₉H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD00053149
CAS Number: 53119-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR