N-Boc-D-prolinal 97%
|
Warning
|
Synonyms:
(R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
, 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-
, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate
, N-Boc-D-prolinal
, (R)-N-BOC-Prolinal
, N-Boc-D-Prolinal
, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester
, N-(tert-Butoxycarbonyl)-D-prolinal
H26294-03
H26294-06
AAH26294-03EA
203.94
USD
AAH26294-03
AAH26294-06
N-Boc-D-prolinal 97%
N-Boc-D-prolinal
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol Boiling Pt: 228 °C Density: 1.06 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD01321389 CAS Number: 73365-02-3 |
Specification Test Results
Optical rotation | +88° (c=1 in chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...