3-Chloro-5-fluoro-4-methoxybenzonitrile 97%
|
Danger
|
H26291-03
H26291-06
AAH26291-06EA
440.67
USD
AAH26291-06
AAH26291-03
3-Chloro-5-fluoro-4-methoxybenzonitrile 97%
3-Chloro-5-fluoro-4-methoxybenzonitrile
Formula:
C₈H₅ClFNO Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115866 CAS Number: 886497-30-9 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...