Dimethyl cyclohexylmalonate 95%

Supplier: Thermo Scientific Chemicals

H26289-06 H26289-03
AAH26289-03EA 89.69 USD
AAH26289-03 AAH26289-06
Dimethyl cyclohexylmalonate 95%
Dimethyl cyclohexylmalonate
Formula: C₁₁H₁₈O₄
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05666206
CAS Number: 49769-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR