2-Butyrylfuran 97%

Warning

H26287-14 H26287-06 H26287-03
AAH26287-14EA 118.46 USD
AAH26287-14 AAH26287-03 AAH26287-06
2-Butyrylfuran 97%
2-Butyrylfuran
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046111
CAS Number: 4208-57-5
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR