3-Fluoro-4-(trifluoromethoxy)benzyl bromide 97%

Danger

H26275-03 H26275-MD
AAH26275-MDEA 114.33 USD
AAH26275-MD AAH26275-03
3-Fluoro-4-(trifluoromethoxy)benzyl bromide 97%
3-Fluoro-4-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD06660205
CAS Number: 886499-04-3
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR