Cyclopropylmagnesium bromide 0.5-0.7 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26273-AC
H26273-AE
AAH26273-AEEA
290.99
USD
AAH26273-AE
AAH26273-AC
Cyclopropylmagnesium bromide 0.5-0.7 M in THF
Cyclopropylmagnesium bromide
Formula:
C₃H₅BrMg Density: 0.968 Flash Pt: -17 °C(1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00672094 CAS Number: 23719-80-4 UN: 3399 ADR: "4.3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...