4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%

Danger

H26246-06 H26246-03
AAH26246-03EA 97.23 USD
AAH26246-03 AAH26246-06
4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%
4-Chloro-3-(trifluoromethylthio)benzyl bromide
Formula: C₈H₅BrClF₃S
MW: 305.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660314
CAS Number: 886503-38-4
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR