3-Chloro-5-(trifluoromethoxy)benzonitrile 97%

Danger

H26239-03 H26239-06
AAH26239-03EA 118.77 USD
AAH26239-03 AAH26239-06
3-Chloro-5-(trifluoromethoxy)benzonitrile 97%
3-Chloro-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD06660309
CAS Number: 886503-29-3
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR