Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-4-cyano-5-(methylthio)thiophene-2-carboxylic acid methyl ester

H26233-09 H26233-03
AAH26233-03EA 50.09 USD
AAH26233-03 AAH26233-09
Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%
Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
Formula: C₈H₈N₂O₂S₂
MW: 228.29 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00052752
CAS Number: 129332-45-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR