2-Methoxy-4-(trifluoromethoxy)benzylamine 97%

Danger

H26231-06 H26231-03
AAH26231-03EA 125.43 USD
AAH26231-03 AAH26231-06
2-Methoxy-4-(trifluoromethoxy)benzylamine 97%
2-Methoxy-4-(trifluoromethoxy)benzylamine
Formula: C₉H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD06660330
CAS Number: 886761-62-2
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR