Vinylcyclopentane 99%

Supplier: Thermo Scientific Chemicals
Danger

H26230-03
AAH26230-03EA 71.99 USD
AAH26230-03
Vinylcyclopentane 99%
Vinylcyclopentane
Formula: C₇H₁₂
Boiling Pt: 97 °C
Density: 0.704
Flash Pt: 10 °C(50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001386
CAS Number: 3742-34-5
EINECS: 223-133-8
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR