2-Methoxy-5-(trifluoromethoxy)benzyl bromide 97%

Danger

H26228-09 H26228-03
AAH26228-09EA 574.98 USD
AAH26228-09 AAH26228-03
2-Methoxy-5-(trifluoromethoxy)benzyl bromide 97%
2-Methoxy-5-(trifluoromethoxy)benzyl bromide
Formula: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04115961
CAS Number: 478484-53-6
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR