4-Nitro-1H-pyrazole-3-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitropyrazole-3-carboxylic acid4-Nitro-3-pyrazolecarboxylic acid

H26224-09
AAH26224-09EA 143.99 USD
AAH26224-09
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1H-pyrazole-3-carboxylic acid
Formula: C₄H₃N₃O₄
MW: 157.085 g/mol
Melting Pt: ca 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00090899
CAS Number: 5334-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR