5-(4-Fluorophenyl)isoxazole 99%
H26188-03
AAH26188-03EA
39.39
USD
AAH26188-03
5-(4-Fluorophenyl)isoxazole 99%
5-(4-Fluorophenyl)isoxazole
Formula:
C₉H₆FNO Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD02183544 CAS Number: 138716-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...