3-Hydroxy-5-(trifluoromethyl)benzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26157-03
AAH26157-03EA
258.99
USD
AAH26157-03
3-Hydroxy-5-(trifluoromethyl)benzoic acid 99%
3-Hydroxy-5-(trifluoromethyl)benzoic acid
Formula:
C₈H₅F₃O₃ MW: 206.12 g/mol Melting Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368783 CAS Number: 328-69-8 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...