4,5-Difluoro-2-hydroxybenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26155-MD
H26155-03
AAH26155-03EA
324.99
USD
AAH26155-03
AAH26155-MD
4,5-Difluoro-2-hydroxybenzonitrile 99%
4,5-Difluoro-2-hydroxybenzonitrile
Formula:
C₇H₃F₂NO MW: 155.10 g/mol Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275287 CAS Number: 186590-36-3 EINECS: None UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...