3-Bromo-5-fluoro-4-methoxyaniline 98%

Warning

H26152-MD H26152-03
AAH26152-MDEA 87.35 USD
AAH26152-MD AAH26152-03
3-Bromo-5-fluoro-4-methoxyaniline 98%
3-Bromo-5-fluoro-4-methoxyaniline
Formula: C₇H₇BrFNO
MW: 220.04 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD08062384
CAS Number: 875664-44-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR