4,5-Difluoro-o-toluidine 99%

Warning

Synonyms: 4,5-Difluoro-2-methylaniline

H26142-03 H26142-MD
AAH26142-MDEA 73.49 USD
AAH26142-MD AAH26142-03
4,5-Difluoro-o-toluidine 99%
4,5-Difluoro-o-toluidine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08062378
CAS Number: 875664-57-6
EINECS: None
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR