4-Hydroxy-2-(trifluoromethyl)benzoic acid 98%

Warning

H26127-03 H26127-MD
AAH26127-MDEA 55.48 USD
AAH26127-MD AAH26127-03
4-Hydroxy-2-(trifluoromethyl)benzoic acid 98%
4-Hydroxy-2-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD01091007
CAS Number: 320-32-1
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR