(5-Fluoro-2-nitrophenyl)acetic acid 97%
|
Warning
|
Synonyms:
5-Fluoro-2-nitrophenylacetic acid
H26126-03
AAH26126-03EA
101.56
USD
AAH26126-03
(5-Fluoro-2-nitrophenyl)acetic acid 97%
(5-Fluoro-2-nitrophenyl)acetic acid
Formula:
C₈H₆FNO₄ MW: 199.14 g/mol Melting Pt: 153…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD07787601 CAS Number: 29640-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...