5'-Fluoro-2'-methoxyacetophenone 99%
|
Warning
|
Synonyms:
1-(5-Fluoro-2-methoxyphenyl)ethanone
H26108-03
AAH26108-03EA
72.76
USD
AAH26108-03
5'-Fluoro-2'-methoxyacetophenone 99%
5'-Fluoro-2'-methoxyacetophenone
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Density: 1.188 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671763 CAS Number: 445-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...