2-Methoxy-5-(trifluoromethoxy)benzaldehyde 98%

Warning

Synonyms: 2-Formyl-4-(trifluoromethoxy)anisole

H26105-03 H26105-MD
AAH26105-MDEA 37.39 USD
AAH26105-MD AAH26105-03
2-Methoxy-5-(trifluoromethoxy)benzaldehyde 98%
2-Methoxy-5-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 20…25 °C
Density: 1.36
Storage Temperature: Ambient
MDL Number: MFCD04115958
CAS Number: 145742-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR