4-Hydroxy-3-(trifluoromethyl)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H26097-03
AAH26097-03EA 254.99 USD
AAH26097-03
4-Hydroxy-3-(trifluoromethyl)benzoic acid 98%
4-Hydroxy-3-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD01091008
CAS Number: 220239-68-9
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR