Benzamidoxime 95%

Danger

Synonyms: Benzamide oximeN'-Hydroxybenzimidamide

H26068-03 H26068-06
AAH26068-06EA 243.08 USD
AAH26068-06 AAH26068-03
Benzamidoxime 95%
Benzamidoxime
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00031485
CAS Number: 613-92-3
EINECS: 210-361-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR