1-(Trifluoromethylcyclopropyl)benzene 97%

Warning

H26056-MD
AAH26056-MDEA 101.78 USD
AAH26056-MD
1-(Trifluoromethylcyclopropyl)benzene 97%
1-(Trifluoromethylcyclopropyl)benzene
Formula: C₁₀H₉F₃
MW: 186.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059543
CAS Number: 883547-73-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR