5-Amino-2-(methylthio)benzotrifluoride 97%

Warning

Synonyms: 4-Methylthio-3-(trifluoromethyl)aniline

H26040-06 H26040-03
AAH26040-03EA 125.43 USD
AAH26040-03 AAH26040-06
5-Amino-2-(methylthio)benzotrifluoride 97%
5-Amino-2-(methylthio)benzotrifluoride
Formula: C₈H₈F₃NS
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD04972923
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR