4-Methyl-3-nitrobenzenesulfonamide 97%
|
Warning
|
H26039-03
AAH26039-03EA
96.41
USD
AAH26039-03
4-Methyl-3-nitrobenzenesulfonamide 97%
4-Methyl-3-nitrobenzenesulfonamide
Formula:
C₇H₈N₂O₄S MW: 216.22 g/mol Melting Pt: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460611 CAS Number: 6949-23-1 EINECS: 230-117-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...