(4-Methylphenyl)methanesulfonyl chloride 97%

Danger

Synonyms: p-Tolylmethanesulfonyl chloride4-Methylbenzylsulfonyl chloridep-Tolylmethanesulphonyl chlorideα-p-Xylenesulfonyl chloride4-Methylbenzylsulphonyl chlorideα-p-Xylenesulphonyl chloride

H26038-03
AAH26038-03EA 80.65 USD
AAH26038-03
(4-Methylphenyl)methanesulfonyl chloride 97%
(4-Methylphenyl)methanesulphonyl chloride
Formula: C₈H₉ClO₂S
MW: 204.67 g/mol
Melting Pt: 75…81 °C
Storage Temperature: Ambient
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR