2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile 95%

Warning

H26022-MD H26022-03
AAH26022-MDEA 61.52 USD
AAH26022-MD AAH26022-03
2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile 95%
2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile
Formula: C₁₁H₁₀ClNO₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD08445711
CAS Number: 441060-95-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR