N-Cbz-4-hydroxycyclohexane predominantly trans 97%

Warning

H26016-06 H26016-14
AAH26016-14EA 301.92 USD
AAH26016-14 AAH26016-06
N-Cbz-4-hydroxycyclohexane predominantly trans 97%
N-Cbz-4-hydroxycyclohexane
Formula: C₁₄H₁₉NO₃
Melting Pt: 162…164 °C
Storage Temperature: Ambient
MDL Number: MFCD02728352
CAS Number: 16801-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR