4-Bromo-2,5-difluorobenzenesulfonamide 98+%
|
Warning
|
H26010-03
AAH26010-03EA
591.44
USD
AAH26010-03
4-Bromo-2,5-difluorobenzenesulfonamide 98+%
4-Bromo-2,5-difluorobenzenesulphonamide
Formula:
C₆H₄BrF₂NO₂S Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569463 CAS Number: 214209-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...