2-Aminoimidazole hemisulfate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminoimidazole sulphate
, 2-Aminoimidazole sulfate
, 1H-imidazol-2-amine sulfate(2:1)
, 1H-imidazol-2-amine sulphate(2:1)
H25999-03
H25999-09
AAH25999-09EA
290.99
USD
AAH25999-09
AAH25999-03
2-Aminoimidazole hemisulfate 98%
2-Aminoimidazole hemisulphate
Formula:
C₆H₁₂N₆O₄S MW: 264,26 g/mol Melting Pt: ca 270 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013162 CAS Number: 1450-93-7 EINECS: 215-918-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...