5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

H25946-03 H25946-09
AAH25946-09EA 338.99 USD
AAH25946-09 AAH25946-03
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin
Formula: C₄₈H₃₈N₄O₄
MW: 734.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012072
CAS Number: 22112-78-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR