Chlorobis(2-methylphenyl)phosphine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-o-tolylchlorophosphine
, Chlorodi(o-tolyl)phosphine
H25890-06
AAH25890-06EA
296.99
USD
AAH25890-06
Chlorobis(2-methylphenyl)phosphine 98%
Chlorobis(2-methylphenyl)phosphine
Formula:
C₁₄H₁₄ClP MW: 248.688 g/mol Boiling Pt: 174…178 °C/3 mm Melting Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038733 CAS Number: 36042-94-1 EINECS: 000-000-0 UN: 1760 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...