2-(Trifluoromethyl)pyrrolidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Trifluoromethyl)tetrahydropyrrole
, 1-Aza-2-(trifluoromethyl)cyclopentane
H25852-03
H25852-MD
AAH25852-MDEA
99.49
USD
AAH25852-MD
AAH25852-03
2-(Trifluoromethyl)pyrrolidine 95%
2-(Trifluoromethyl)pyrrolidine
Formula:
C₅H₈F₃N MW: 139.12 g/mol Boiling Pt: 110…112 °C Density: 1.21 Flash Pt: 26 °C(78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02663405 CAS Number: 109074-67-1 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...