4-(4-Fluorobenzoyl)butyric acid 98%

Warning

Synonyms: 5-(4-Fluorophenyl)-5-oxovaleric Acid5-(4-Fluorophenyl)-5-oxopentanoic acid

H25839-03 H25839-06
AAH25839-06EA 261.96 USD
AAH25839-06 AAH25839-03
4-(4-Fluorobenzoyl)butyric acid 98%
4-(4-Fluorobenzoyl)butyric acid
Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00667222
CAS Number: 149437-76-3
EINECS: 604-692-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR