5-Bromo-4,6-dichloropyrimidine 97%
|
Danger
|
H25821-06
H25821-03
AAH25821-06EA
447.02
USD
AAH25821-06
AAH25821-03
5-Bromo-4,6-dichloropyrimidine 97%
5-Bromo-4,6-dichloropyrimidine
Formula:
C₄HBrCl₂N₂ Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047379 CAS Number: 68797-61-5 EINECS: 272-305-9 UN: 1759 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...