1-Benzhydryl-3-azetidinol hydrochloride 95%

Warning

Synonyms: 1-(Diphenylmethyl)-3-hydroxyazetidine hydrochloride1-Benzhydrylazetidin-3-ol hydrochloride

H25819-03 H25819-06
AAH25819-06EA 103.54 USD
AAH25819-06 AAH25819-03
1-Benzhydryl-3-azetidinol hydrochloride 95%
1-Benzhydryl-3-azetidinol hydrochloride
Formula: C₁₆H₁₇NO·HCl
MW: 275.78 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00209447
CAS Number: 90604-02-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR