1H-Pyrazole-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Pyrazolecarboxylic acid

H25783-06 H25783-03
AAH25783-03EA 255.99 USD
AAH25783-03 AAH25783-06
1H-Pyrazole-4-carboxylic acid 97%
1H-Pyrazole-4-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 281…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00011558
CAS Number: 37718-11-9
EINECS: 253-641-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR