3,5-Dinitro-4-methylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methyl-3,5-dinitrobenzeneboronic acid
H25773-14
H25773-06
H25773-03
AAH25773-14EA
1577.99
USD
AAH25773-14
AAH25773-03
AAH25773-06
3,5-Dinitro-4-methylphenylboronic acid 97%
3,5-Dinitro-4-methylphenylboronic acid
Formula:
C₇H₇BN₂O₆ Melting Pt: ca 280 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD08060935 CAS Number: 28249-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...