Ethyl-5-amino-1,2,3-thiadiazole-4-carboxylate ≥98%

Warning

H25762-03
AAH25762-03EA 451.52 USD
AAH25762-03
Ethyl-5-amino-1,2,3-thiadiazole-4-carboxylate ≥98%
Ethyl-5-amino-1,2,3-thiadiazole-4-carboxylate
Formula: C₅H₇N₃O₂S
MW: 173.19 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00778317
CAS Number: 6440-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR