Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 96%

Supplier: Thermo Scientific Chemicals
Warning

H25757-06 H25757-03 H25757-MD H25757-03 H25757-06
AAH25757-03EA 267.99 USD
CAAAH25757-03 CAAAH25757-06 AAH25757-03 AAH25757-MD AAH25757-06
Methyl-1-trifluoromethyl-1-cyclopropanecarboxylate 96%
Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
Formula: C₆H₇F₃O₂
MW: 168.12
Boiling Pt: 115 °C
Flash Pt: 42 °C(108 °F)
Storage Temperature: Ambient
MDL Number: MFCD06801321
CAS Number: 208242-25-5
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR