2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine 97%

Supplier: Thermo Scientific Chemicals

Warning

B25742-03
AAB25742-03EA 102 USD
AAB25742-03
2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine 97%
2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine
Formula: C₁₉H₂₀N₆O
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00272362
CAS Number: 43167-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR