1H-Pyrazole-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Pyrazolecarboxylic acid
B25732-03
B25732-06
AAB25732-06EA
200.99
USD
AAB25732-06
AAB25732-03
1H-Pyrazole-3-carboxylic acid 97%
1H-Pyrazole-3-carboxylic acid
Formula:
C₄H₄N₂O₂ MW: 112.09 g/mol Melting Pt: 211 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00077436 CAS Number: 1621-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...