3-Pyridineacetamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Pyridin-3-yl)acetamide
B25692-14
B25692-06
AAB25692-14EA
166.99
USD
AAB25692-14
AAB25692-06
3-Pyridineacetamide 99%
3-Pyridineacetamide
Formula:
C₇H₈N₂O MW: 136.15 g/mol Melting Pt: 122…124 °C Density: 1,17 Storage Temperature: Ambient |
MDL Number:
MFCD08236789 CAS Number: 3724-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...