1-Bromo-2-isopropoxybenzene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2'-Bromophenoxy)propane

B25688-06 B25688-14
AAB25688-06EA 65.89 USD
AAB25688-06 AAB25688-14
1-Bromo-2-isopropoxybenzene 97%
1-Bromo-2-isopropoxybenzene
Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 73 °C/2 mm
Density: 1.34
Storage Temperature: Ambient
MDL Number: MFCD00070759
CAS Number: 701-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR