2,5-Dichlorobenzenesulfonic acid dihydrate 97%

Danger

B25650-14 B25650-22
AAB25650-14EA 72.32 USD
AAB25650-14 AAB25650-22
2,5-Dichlorobenzenesulfonic acid dihydrate 97%
2,5-Dichlorobenzenesulphonic acid dihydrate
Formula: C₆H₄Cl₂O₃S·2H₂O
MW: 263.09 g/mol (227.07 anhy)
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD08271971
CAS Number: 38484-94-5
EINECS: 201-829-2
UN: 2585
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR